Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Jars with snap-on lid

Jars with snap-on lid

5600 ml, 222 mm, 190 mm
Catalog # SEMA3059
Supplier:   SEMADENI
Jars with snap-on lid
Catalog # SEMA3059
Supplier:  SEMADENI
Jars with snap-on lid
Jars with snap-on lid

5600 ml, 222 mm, 190 mm

Price
SFr. 302.00
Log in for availability
1 * 100 ST
Product Details & Documents

Jars, made of PP, with close-fitting snap-on lid.

  • Good temperature resistance
  • Good chemical resistance

 : *The product(s) on this page are outside the scope of the EU Directive 2019/904

Specifications
  • Ext.Ø:
    222 mm
  • Capacity:
    5600 ml
  • Height:
    190 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.