Order Entry
Switzerland
ContactUsLinkComponent
Jars with screw cap
Jars with screw cap
Catalog # SEMA3046
Supplier:  SEMADENI
undefined
Jars with screw cap
Catalog # SEMA3046
Supplier:  SEMADENI

Specifications

  • Ext.Ø:
    55 mm
  • Capacity:
    50 ml
  • Height:
    45 mm
  • Pk:
    100

Specifications

About this item

Jars, made of PP, with double walled bottom and tri-seal insert in the lid.