Order Entry
Switzerland
ContactUsLinkComponent
 
Jars, wide neck, with screw cap
Catalog # 216-3149
Supplier:  SEMADENI
undefined
Jars, wide neck, with screw cap
Catalog # 216-3149
Supplier:  SEMADENI

Specifications

  • Ext.Ø:
    90 mm
  • Capacity:
    500 ml
  • Height:
    100 mm
  • Pk:
    1

Specifications

About this item

Jars, made of PP, translucent, with screw cap.