ContactUsLinkComponent
Büchner funnels, PYREX®
  CORN36060-30C
 :  Corning
undefined
Büchner funnels, PYREX®
  CORN36060-30C
 :  Corning
  • Plate Ø:
    30 mm
  • Porosity:
    Coarse
  • Capacity:
    30 ml
  • Seq:
    03
  • Pk:
    1
PYREX®, borosilicate glass, with sintered glass disc.
  • No filter papers required
  • Highly resistant to most corrosive reagents including ammonia and concentrated sulphuric acid
  • Very good temperature resistance
Especially useful where filter paper would be attacked by material being filtered. Top edge is beaded.
 : Disc should not be subjected to pressure differential in excess of 1,01 bar.