ContactUsLinkComponent
Beakers, low form, borosilicate glass, heavy-duty, double scale, KIMAX®, KIMBLE
Catalog # SCERSP14005-1000
Anbieter:  DWK Life Sciences
Supplier #: SP14005-1000
undefined
Beakers, low form, borosilicate glass, heavy-duty, double scale, KIMAX®, KIMBLE
Catalog # SCERSP14005-1000
Anbieter:  DWK Life Sciences
Supplier #: SP14005-1000
Anmelden für Verfügbarkeit
Select an Option
107 mm
Select an Option
50 ml
Select an Option
1000 ml
Select an Option
152 mm
Select an Option
04
Select an Option
24
Über diesen Artikel
Manufactured from 33 expansion, low extractable borosilicate glass. Thick uniform walls throughout and extra wall thickness built into the evenly tooled top rim.
  • Uniformity of construction allows for use on hot plates
  • All sizes have a durable matte finish marking area and a white graduated scale
  • Design meets ASTM specification E960, type II requirements
Heavy-duty glass 'thick wall' beakers offer superior mechanical strength and durability. Improved safety when used under extreme conditions such as mechanized washing and rough handling.
Certifications: Conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Frequently Bought Together