Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ammonium molybdate solutions
Ammonium molybdate solution R3 Reag. Ph. Eur. 1005704
Ammonium molybdate solution R3 Reag. Ph. Eur. 1005704
Corrosive
Ammonium molybdate solution R3 Reag. Ph. Eur. 1005704
Catalog # 87772.290
Supplier:
VWR Chemicals
CAS Number:
SDS
Certificates
Expand image
Corrosive
Ammonium molybdate solution R3 Reag. Ph. Eur. 1005704
Catalog # 87772.290
Supplier:
VWR Chemicals
CAS Number:
SDS
Certificates
Specifications
Pack type:
Plastic bottle
Pk:
1 L
Seq:
5037
Specifications
About this item
Product Details
Documentation
:
UN:
1760
ADR:
8,I
:
GHS05
Product Details
Documentation