Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ferric ammonium sulphate solution
Ferric ammonium sulphate solution R5 Reag. Ph. Eur. 1037704
Ferric ammonium sulphate solution R5 Reag. Ph. Eur. 1037704
87831.180
:
VWR Chemicals
:
Ferric ammonium sulphate solution R5 Reag. Ph. Eur. 1037704
87831.180
:
VWR Chemicals
:
Pack type:
Plastic bottle
Pk:
100 mL
Seq:
2440
:
:
1760
:
8,I
:
GHS05