Order Entry
Switzerland
ContactUsLinkComponent
Ammonium iron(II) sulphate 100 g/l solution R2 Reag. Ph. Eur. 1037702
Ammonium iron(II) sulphate 100 g/l solution R2 Reag. Ph. Eur. 1037702
Catalog # 87830.290
Supplier:  VWR Chemicals
CAS Number:  
undefined
Ammonium iron(II) sulphate 100 g/l solution R2 Reag. Ph. Eur. 1037702
Catalog # 87830.290
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :284.05 g/mol
Storage Temperature:Ambient
UN:3264
ADR:8,II
CAS Number:10045-89-3
Formula:H₈FeN₂O₈S₂
MDL Number:MFCD00051039