ContactUsLinkComponent
Racks for Stomacher® blender bags
Racks for Stomacher® blender bags
Catalog # 432-3581
Supplier:  DELTALAB
Racks for Stomacher® blender bags
Catalog # 432-3581
Supplier:  DELTALAB
Specifications
  • Compartments:
    14
  • Description:
    Rack for 14 Stomacher® bags, stainless steel
  • W×D×H:
    390×200×240 mm
  • Pk:
    1
Specifications
About this item
These racks are made of stainless steel wire with side handles incorporated.
  • Very good chemical resistance
  • Very good temperature resistance
  • Sturdy