Orders LinkContactUsLinkComponent
Jars with screw cap
Jars with screw cap
Supplier:   SEMADENI
Jars with screw cap
Supplier:  SEMADENI
Certificates
Jars with screw cap
Jars with screw cap
4 Options Available Below
Product Details & Documents
Jars made of PP, white,with screw caps made of red PP.
  • Very good chemical resistance
  • Very good temperature resistance
Suitable for powders, granules, pastes or cosmetic products. Physiologically harmless and suitable for food storage.
Product Family Options
Product Information
  • Capacity
    Ext.Ø
    Height
    Availability
    Price
  • 60 ml
    45 mm
    55 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - SFr. 55.00
    SFr. 55.00
    1 * 100 ST
       SFr. 55.00
  • 125 ml
    56 mm
    67 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - SFr. 66.20
    SFr. 66.20
    1 * 100 ST
       SFr. 66.20
  • 250 ml
    73 mm
    78 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - SFr. 99.10
    SFr. 99.10
    1 * 100 ST
       SFr. 99.10
  • 1000 ml
    113 mm
    128 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - SFr. 286.00
    SFr. 286.00
    1 * 100 ST
       SFr. 286.00