Order Entry
Switzerland
ContactUsLinkComponent
Beakers, tall form, borosilicate glass
Beakers, tall form, borosilicate glass
Catalog # KAVA632427012250
Supplier:  Kavalier
undefined
Beakers, tall form, borosilicate glass
Catalog # KAVA632427012250
Supplier:  Kavalier

Specifications

  • Ext.Ø:
    60 mm
  • Capacity:
    250 ml
  • Height:
    120 mm
  • Pk:
    1

Specifications

About this item

Borosilicate glass 3,3.

  • Very good chemical resistance
  • High temperature resistance
  • Autoclavable

Ideal for general laboratory use. With spout and white graduations.