Orders LinkContactUsLinkComponent
Jars, with screw cap
Jars, with screw cap
Supplier:   SEMADENI
Jars, with screw cap
Supplier:  SEMADENI
Certificates
Jars, with screw cap
Jars, with screw cap
6 Options Available Below
Product Details & Documents
PP, white.
  • Good chemical resistance
  • Good temperature resistance
Ideal for collecting, storing and transporting samples.
Product Family Options
Product Information
  • Capacity
    Ext.Ø
    Height
    Colour
    Availability
    Price
  • 6 ml
    28 mm
    18 mm
    White
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - SFr. 20.60
    SFr. 20.60
    1 * 100 ST
       SFr. 20.60
  • 25 ml
    34 mm
    40 mm
    White
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - SFr. 27.50
    SFr. 27.50
    1 * 100 ST
       SFr. 27.50
  • 35 ml
    38 mm
    48 mm
    White
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - SFr. 33.70
    SFr. 33.70
    1 * 100 ST
       SFr. 33.70
  • 60 ml
    45 mm
    55 mm
    White
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - SFr. 70.00
    SFr. 70.00
    1 * 100 ST
       SFr. 70.00
  • 125 ml
    56 mm
    67 mm
    White
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - SFr. 87.40
    SFr. 87.40
    1 * 100 ST
       SFr. 87.40
  • 500 ml
    95 mm
    91 mm
    White
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - SFr. 201.00
    SFr. 201.00
    1 * 100 ST
       SFr. 201.00