Flasks, round bottom, with two or three necks and standard ground joints
Supplier: Kavalier
Certificates
About this item
Borosilicate glass 3,3.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Typical applications: distillation, extraction.
Frequently Bought Together