Cell scrapers
Marketsource
Supplier: BELLCO GLASS
Jump to:
About this item
Stainless steel blade holder mounted on plastic covered stainless steel handle.
3 Options Available Below
Product Details & Documents
Stainless steel blade holder mounted on plastic covered stainless steel handle.
Product Family Options
Product Information
- AvailabilityPrice
- BELC7731-22000Small cell scraper, stainless steel, 15,88×6,35 mm blade, handle length 197 mmSmall cell scraper, stainless steel, 15,88×6,35 mm blade, handle length 197 mmLog in for availability1 * 2 STSFr. 123.92Specifications:MarketsourceMarketsource
- BELC7731-22005Cell scraper, Medium, replacement blade, Stainless steel, Blade width (mm): 31,75 mmCell scraper, Medium, replacement blade, Stainless steel, Blade width (mm): 31,75 mmLog in for availability1 * 10 STSFr. 131.92Specifications:MarketsourceMarketsource
- BELC7731-22115Replacement blade, stainless steel, medium, 31,75×14,29 mmReplacement blade, stainless steel, medium, 31,75×14,29 mmLog in for availability1 * 10 STSFr. 45.03Specifications:MarketsourceMarketsource
Consigli
I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".
Altrimenti riceverai suggerimenti generici..