Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Storage Boxes with Lid

Storage Boxes with Lid

Supplier:   SEMADENI
Storage Boxes with Lid
Supplier:  SEMADENI
Certificates
Storage Boxes with Lid
Storage Boxes with Lid

3 Options Available Below

Product Details & Documents

These storage boxes with transparent lids are either made of polystyrene (PS) or styrene-acrylonitrile resin (SAN).

Product Family Options

Product Information

  • Capacity
    Colour
    W×D×H
    Material
    Availability
    Price
  • 0,5 l
    Transparent
    100×100×85 mm
    PS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 11.20
    SFr. 11.20
    1 * 1 ST
       SFr. 11.20

  • 1,5 l
    Transparent
    200×100×85 mm
    PS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 10.00
    SFr. 10.00
    1 * 1 ST
       SFr. 10.00

  • 6 l
    Transparent
    340×230×95 mm
    SAN
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 66.20
    SFr. 66.20
    1 * 1 ST
       SFr. 66.20