Ball Valve for Storage Bottles
Ball valve, PP, white, ½" NW 16
Catalog # 229-7489
Supplier: SEMADENI
Jump to:
Product Details & Documents
½” universal valve made from PP, resistant to acids, alkalis and solvents.
- Fully sealed design, runs very easily and with no blockages
- Outlet spout Ø 12 mm
Housing and ball made from PP, ball seal made from PTFE and O ring made from nitrile rubber (NBR 70).
Specifications
- Port Size:½" NW 16
- Description:Ball valve, PP, white