Orders LinkContactUsLinkComponent
Humidity standards
Humidity standards
Humidity standards
Certificates
Humidity standards
Humidity standards
8 Options Available Below
Product Details & Documents
Although most relative humidity (RH) probes have excellent long-term stability, it is recommended that their calibration be checked regularly.
  • Traceable to national standard – SCS (Swiss Calibration Service) certificate included
  • Ampoules contain unsaturated salt solutions (~0,8 ml)
  • Inexpensive on-site calibration
  • Simple and safe to use
  • 10 year shelf life
  • Practical packs of five ampoules of the same humidity value
Delivery information: Each ampoule is labelled with the humidity value and serial number.
Caution: Values and accuracy (measurement uncertainty values) below are measured at 23 °C.
Product Family Options
Product Information
  • Accuracy
    Relative humidity (RH%)
    Pk
    Availability
    Price
  • 620-2272
    Certified humidity standards
    Certified humidity standards
    ±0,3% RH
    0,5%
    5 Ampoul
    Log in for availability
    Log in for availability
    1 * 5 Ampull
    1 * 5 Ampull - SFr. 91.20
    SFr. 91.20
    1 * 5 Ampull
       SFr. 91.20
  • 620-2223
    Certified humidity standards
    Certified humidity standards
    ±0,3% RH
    10%
    5 Ampoul
    Log in for availability
    Log in for availability
    1 * 5 Ampull
    1 * 5 Ampull - SFr. 91.20
    SFr. 91.20
    1 * 5 Ampull
       SFr. 91.20
  • 620-2758
    Certified humidity standards
    Certified humidity standards
    ±0,3% RH
    20%
    5 Ampoul
    Log in for availability
    Log in for availability
    1 * 5 Ampull
    1 * 5 Ampull - SFr. 91.20
    SFr. 91.20
    1 * 5 Ampull
       SFr. 91.20
  • 620-2224
    Certified humidity standards
    Certified humidity standards
    ±0,5% RH
    35%
    5 Ampoul
    Log in for availability
    Log in for availability
    1 * 5 Ampull
    1 * 5 Ampull - SFr. 91.20
    SFr. 91.20
    1 * 5 Ampull
       SFr. 91.20
  • 620-2225
    Certified humidity standards
    Certified humidity standards
    ±0,9% RH
    50%
    5 Ampoul
    Log in for availability
    Log in for availability
    1 * 5 Ampull
    1 * 5 Ampull - SFr. 91.20
    SFr. 91.20
    1 * 5 Ampull
       SFr. 91.20
  • 707-0044
    Certified humidity standards
    Certified humidity standards
    ±0,9% RH
    65%
    5 Ampoul
    Log in for availability
    Log in for availability
    1 * 5 Ampull
    1 * 5 Ampull - SFr. 91.20
    SFr. 91.20
    1 * 5 Ampull
       SFr. 91.20
  • 620-2226
    Certified humidity standards
    Certified humidity standards
    ±1,2% RH
    80%
    5 Ampoul
    Log in for availability
    Log in for availability
    1 * 5 Ampull
    1 * 5 Ampull - SFr. 91.20
    SFr. 91.20
    1 * 5 Ampull
       SFr. 91.20
  • 707-0045
    Certified humidity standards
    Certified humidity standards
    ±1,2% RH
    95%
    5 Ampoul
    Log in for availability
    Log in for availability
    1 * 5 Ampull
    1 * 5 Ampull - SFr. 91.20
    SFr. 91.20
    1 * 5 Ampull
       SFr. 91.20