ContactUsLinkComponent
Beakers, low form, stainless steel
  213-1802
 :  RSG
 : 075.250
undefined
Beakers, low form, stainless steel
  213-1802
 :  RSG
 : 075.250
Select an Option
65 mm
Select an Option
250 ml
Select an Option
80 mm
Select an Option
02
Select an Option
1
Stainless steel 18/10, with spout, straight edge.
  • Good chemical resistance
  • Excellent resistance to heat
  • Smooth surfaces for fast, easy cleaning
Ideal for corrosive materials. Robust and durable.
Frequently Bought Together