ContactUsLinkComponent
Arsenic standard solution, 1,000 mg/L As in dil. nitric acid (from As) standard for AAS and ICP
Anbieter:  PerkinElmer
Zertifikate
Arsenic standard solution, 1,000 mg/L As in dil. nitric acid (from As) standard for AAS and ICP
Anbieter:  PerkinElmer
Frequently Bought Together