Order Entry
Switzerland
ContactUsLinkComponent
Vanadium standard solution, 1000 mg/L V in dil. nitric acid (from V) standard for AAS and ICP
Vanadium standard solution, 1000 mg/L V in dil. nitric acid (from V) standard for AAS and ICP
Catalog # PERCN9300165
Supplier:  PerkinElmer
CAS Number:  
Vanadium standard solution, 1000 mg/L V in dil. nitric acid (from V) standard for AAS and ICP
Catalog # PERCN9300165
Supplier:  PerkinElmer
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    2% HNO₃
  • Pk:
    1
  • Element:
    Vanadium
  • Symbol:
    V

Specifications

About this item