Order Entry
Switzerland
ContactUsLinkComponent
Acrylamide ≥98.0% (by GC, titration analysis)
Toxic
Health
Acrylamide ≥98.0% (by GC, titration analysis)
Supplier:  TCI
Certificates
undefined
Toxic
Health
Acrylamide ≥98.0% (by GC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Bromination Method):
    min. 98.0 %
  • Melting point:
    83.0 to 87.0 deg-C
  • Solubility in Water:
    almost transparency

Specification Test Results

Related Information

Frequently Bought Together