Methyl arachidate
Supplier: MATREYA
Jump to:
About this item
Synonyms: Methyl eicosanoate, Methyl Arachidate
- CAS Number:1120-28-1
- Formula:C₂₁H₄₂O₂
- MDL Number:MFCD00009014
- Molecular Weight:326.56 g/mol
- EINECS:214-304-8
1 Options Available Below
Product Details & Documents
Synonyms: Methyl eicosanoate, Methyl Arachidate
- CAS Number:1120-28-1
- Formula:C₂₁H₄₂O₂
- MDL Number:MFCD00009014
- Molecular Weight:326.56 g/mol
- EINECS:214-304-8
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



