Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Methyl arachidate ≥98%

Methyl arachidate ≥98%

Methyl arachidate ≥98%
Certificates
Methyl arachidate ≥98%
Methyl arachidate ≥98%

2 Options Available Below

Product Details & Documents

Purity: 98+% CAS No.: 1120-28-1 Molecular Formula: CH3(CH2)18COOCH3 Formula Weight: 326.56 Flash Point: 110°(230°F) Physical Form: Crystalline powder Melting Point: 48° Boiling Point: 215°/10mm MDL No.: MFCD00009014

Synonyms:  Methyl eicosanoate, Methyl Arachidate
  • CAS Number:1120-28-1
  • Formula:C₂₁H₄₂O₂
  • MDL Number:MFCD00009014
  • Molecular Weight:326.56 g/mol
  • Boiling Pt:215 °C (10 mmHg)
  • Melting Pt:48 °C
  • Flash Pt:>110 °C (230 °F)
  • Storage Temperature:Ambient
  • EINECS:214-304-8
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.