
3,5-Dichloro-2-hydroxybenzenesulphonic acid sodium salt suitable for peroxidase analysis, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Certificates
About this item
CAS Number:54970-72-8
:
Formula:C₆H₃Cl₂NaO₄S
MDL Number:MFCD00009798
:265.05 g/mol
Melting Pt:>300 °C
:M-Clarity™ quality level = MQ200
EINECS:259-416-8
:GHS07
Storage Temperature:Room Temperature
Specification Test Results
- Appearance (Colour):White to Off-White
- Appearance (Form):Powder
- ICP Major Analysis
Confirms Sodium Component:Confirmed - Purity (TLC):≥98 %
- Solubility (Colour):Colorless to Faint Yellow
- Solubility (Turbidity)
5% H2O:Clear to Slightly Hazy - Recommended Retest Period:2 Years
Specification Test Results
Frequently Bought Together