Order Entry
Switzerland
ContactUsLinkComponent
D-(+)-Glucose-[¹³C₆] (99% ¹³C )
D-(+)-Glucose-[¹³C₆] (99% ¹³C )
Catalog # CAMBCLM-1396-50
CAS Number:  
D-(+)-Glucose-[¹³C₆] (99% ¹³C )
Catalog # CAMBCLM-1396-50
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g
  • Seq:
    3368

Specifications

About this item

 :
 :186.110 g/mol
CAS Number:110187-42-3
Formula:C₆H₁₂O₆
MDL Number:MFCD00037544