Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Estradiol

Glass bottle for solids, 5 g
Catalog # MOLEM59540594
Marketsource
Supplier:   Molekula
Estradiol
Catalog # MOLEM59540594
Supplier:  Molekula
Marketsource
CAS Number:  50-28-2
Estradiol

Glass bottle for solids, 5 g

CAS Number:  50-28-2
Price
Inquire For Price
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  beta-Estradiol, 17β-Estradiol
  • CAS Number:50-28-2
  • Formula:C₁₈H₂₄O₂
  • MDL Number:MFCD00003693
  • Molecular Weight:272.39 g/mol
  • EINECS:200-023-8
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.