Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Estradiol

Estradiol

Supplier:   Cayman Chemical
Estradiol
Supplier:  Cayman Chemical
Certificates

4 Options Available Below

Product Details & Documents
Synonyms:  beta-Estradiol, 17β-Estradiol
  • CAS Number:50-28-2
  • Formula:C₁₈H₂₄O₂
  • MDL Number:MFCD00003693
  • Molecular Weight:272.39 g/mol
  • EINECS:200-023-8
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    500 mg
    Log in for availability
    Log in for availability
    1 * 500 mg
    1 * 500 mg - SFr. 49.30
    SFr. 49.30
    1 * 500 mg
       SFr. 49.30

  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - SFr. 93.10
    SFr. 93.10
    1 * 1 g
       SFr. 93.10

  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - SFr. 289.00
    SFr. 289.00
    1 * 5 g
       SFr. 289.00

  • Glass bottle for solids
    10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g - SFr. 385.00
    SFr. 385.00
    1 * 10 g
       SFr. 385.00

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.