4'-Methoxyacetoacetanilide
Catalog # ACTIAS40683G5
Supplier: ACTIVATE SCIENTIFIC
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:207.2300 g/mol
EINECS:226-615-6
CAS Number:5437-98-9
Formula:C₁₁H₁₃NO₃
MDL Number:MFCD00008783