Order Entry
Switzerland
Orders LinkContactUsLinkComponent
4-(Methylthio)benzylamine

4-(Methylthio)benzylamine

Glass bottle for solids, 5 g
Catalog # WAKO358-14563
Marketsource
Supplier:   Wako
4-(Methylthio)benzylamine
Catalog # WAKO358-14563
Supplier:  Wako
Marketsource
CAS Number:  83171-39-5
4-(Methylthio)benzylamine

Glass bottle for solids, 5 g

CAS Number:  83171-39-5
Price
Inquire For Price
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  (4-(Methylthio)phenyl)methanamine
  • CAS Number:83171-39-5
  • Formula:C₈H₁₁NS
  • MDL Number:MFCD01310831
  • Molecular Weight:153.2500 g/mol
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.