Bottles, wide neck, octagonal, with standard ground stoppers
: Kavalier
Soda-glass, clear or amber.
- Good chemical and physical properties
- For applications that do not involve high thermal stresses
Ideal for sampling, storing and transporting powders and pastes. Amber coloured to protect light-sensitive materials.
Frequently Bought Together