Orders LinkContactUsLinkComponent
Dosing bottles, LDPE
Dosing bottles, LDPE
Supplier:   SEMADENI
Dosing bottles, LDPE
Supplier:  SEMADENI
Certificates
Dosing bottles, LDPE
Dosing bottles, LDPE
2 Options Available Below
Product Details & Documents
Wide neck translucent LDPE dosing bottles with white measuring cap and are suitable for viscous liquids.
  • Dose opening of Ø 5 mm
  • Neck Ø 44 mm
  • Good chemical and temperature resistance
Product Family Options
Product Information
  • Capacity
    Height
    Ext.Ø
    Availability
    Price
  • 250 ml
    152 mm
    62 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 2.80
    SFr. 2.80
    1 * 1 ST
       SFr. 2.80
  • 1000 ml
    152 mm
    85 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 4.00
    SFr. 4.00
    1 * 1 ST
       SFr. 4.00