Dosing bottle, HDPE
100 ml, 115 mm, 41 mm
215-0241
: SEMADENI
Natural or translucent cylindrical shaped dosage bottle with hinged cap, protecting the jet orifice of Ø 3 mm.
- For easy handling and flow dosing
- Waterproof bottle when cap is closed
- Good chemical and temperature resistance
- Ext.Ø:41 mm
- Capacity:100 ml
- Height:115 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



