Inverted fluorescence microscopes, Axio Vert A1 FL-LED
: ZEISS
The Axio Vert A1 FL-LED has a four-position mount for LED modules for reflected light fluorescence. One position is for green fluorescent protein (GFP).
- Differential interference contrast to observe the finest cells structures (optional)
- LED fluorescence, long lifetime, homogeneous illumination with full intensity
- No heating and cooling period required during the LED fluorescence
- Ergonomic design to comfortably work in an upright position
- Optional contrast systems (iHMC, PlasDIC, DIC)
- Catalog No:
- ZEIS4880019901000
- ZEIS4230529572000
- ZEIS4230529511000
- ZEIS4278099000000
- 630-2152
- ZEIS4880140000000
- ZEIS4230529552000
- ZEIS4880499901400
- ZEIS0000001374896
- ZEIS4880509901000
- Eyepieces:
- WF 10×/23 mm
- Nosepiece:
- Quintuple
- Illumination:
- LED fluorescence
- Electrical:
- 12 V DC 60 W; 100 – 240 V/AC; 50 – 60 Hz