ContactUsLinkComponent
Incinerating dishes, square
  KAVA641322255201
 :  Kavalier
Incinerating dishes, square
  KAVA641322255201
 :  Kavalier
  • Type:
    without spout
  • Capacity:
    13 ml
  • W×D×H:
    62×40×12 mm
  • Environmentally Preferable:
  • Pk:
    1
Porcelain.