Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Bottles
Plastic Bottles
Bottles, wide neck, HDPE, rectangular, with screw cap
Bottles, wide neck, HDPE, rectangular, with screw cap
Bottles, wide neck, HDPE, rectangular, with screw cap
SEMA1830
:
SEMADENI
Bottles, wide neck, HDPE, rectangular, with screw cap
SEMA1830
:
SEMADENI
Capacity:
1000 ml
W×D×H:
87×87×180 mm
Neck I-Ø:
39 mm
Pk:
1
HDPE, translucent.
Good chemical resistance
Good temperature resistance
Tight closure