Order Entry
Switzerland
ContactUsLinkComponent
Drums, UN approved, with screw lids
Drums, UN approved, with screw lids
Catalog # SEMA0719
Supplier:  SEMADENI
Drums, UN approved, with screw lids
Catalog # SEMA0719
Supplier:  SEMADENI

Specifications

  • Ext.Ø:
    580 mm
  • Capacity:
    220 L
  • Height:
    940 mm
  • Pk:
    1

Specifications

About this item

Drums, made of blue HDPE.

  • Very sturdy
  • Good chemical resistance
  • Good temperature resistance
  • UN-approved