Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Buckets, Round

Natural, 17 L, 330 mm, 330 mm
Catalog # 215-0274
Supplier:   SEMADENI
Buckets, Round
Catalog # 215-0274
Supplier:  SEMADENI
Buckets, Round
Buckets, Round

Natural, 17 L, 330 mm, 330 mm

Price
SFr. 31.70
Log in for availability
1 * 1 ST
Product Details & Documents

Round buckets, made of HDPE, natural, with graduation, handle and spout.

  • Sturdy
  • Good chemical resistance
  • Hot water resistant
Specifications
  • Capacity:
    17 L
  • Colour:
    Natural
  • Height:
    330 mm
  • Ø ext.:
    330 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.