Sleeves for conical ground joints
Katalognr. SEMA0614
Anbieter: SEMADENI
Technische Daten
- For NS:29/32
- Pk:1
Technische Daten
Über diesen Artikel
Made of PTFE, white.
- Very good chemical resistance
- Gas, liquid and vacuum-tight (to 0,1 mbar)
- Temperature resistant from −200 to +280 °C
For fitting onto standard ground cones. Easily detachable and suitable for repeated use. Joint grease is not needed.