Potassium dichromate standardised solution 0.025 N in aqueous solution
Supplier: Thermo Fisher Scientific
Potassium dichromate standardised solution 0.025 N in aqueous solution
Supplier: Thermo Fisher Scientific
Certificates
Jump to:
About this item
CAS No.: 7778-50-9 Molecular Formula: Cr2K2O7 Formula Weight: 294.18 Physical Form: Liquid MDL No.: MFCD00011367
- CAS Number:7778-50-9
- Formula:K₂Cr₂O₇
- MDL Number:MFCD00011367
- Molecular Weight:294.18 g/mol
- Storage Temperature:Ambient
- EINECS:231-906-6
- UN:3287
- ADR:6.1,III
2 Options Available Below
Product Details & Documents
CAS No.: 7778-50-9 Molecular Formula: Cr2K2O7 Formula Weight: 294.18 Physical Form: Liquid MDL No.: MFCD00011367
- CAS Number:7778-50-9
- Formula:K₂Cr₂O₇
- MDL Number:MFCD00011367
- Molecular Weight:294.18 g/mol
- Storage Temperature:Ambient
- EINECS:231-906-6
- UN:3287
- ADR:6.1,III



