Cholecalciferol (Vitamin D3), EMPROVE® ESSENTIAL BP, Ph. Eur., USP suitable for use as excipient, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Certificates
About this item
Synonyms: Vitamin D3, Colecalciferol
CAS Number:67-97-0
:
Formula:C₂₇H₄₄O
MDL Number:MFCD00078131
:384.65 g/mol
Storage Temperature:Refrigerator
EINECS:200-673-2
UN:2811
ADR:6.1,I
:GHS06
:GHS08
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together