
Strontium chloride hexahydrate 99.0-103.0%, crystals, BAKER ANALYZED® ACS low in magnesium, J.T.Baker®
Catalog # 4036-01
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Poly bottle, white, blue T/E cap
- Pk:500 g
- Seq:3645
Specifications
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:99.0 - 103.0 %
- ACS - Assay (SrCl₂·6H₂O):99.0 - 103.0 %
- ACS - Insoluble Matter:≤0.005 %
- ACS - pH of 5% Solution at 25°C:5.0 - 7.0
- ACS - Sulfate (SO₄):≤0.001 %
- ACS - Barium (Ba):≤0.05 %
- ACS - Calcium (Ca):≤0.05 %
- Trace Impurities - ACS - Heavy Metals (as Pb):≤5 ppm
- Trace Impurities - Iron (Fe):≤5 ppm
- Trace Impurities - Magnesium (Mg):≤2.000 ppm
Specification Test Results
About this item
:
:266.62 g/mol
Merck Index:13,08920
Boiling Pt:100 °C (1013 hPa)
Melting Pt:61 °C
Density:1.93 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:233-971-6
:GHS07
CAS Number:10025-70-4
Formula:SrCl₂·6H₂O
MDL Number:MFCD00149865