Buffer, sodium acetate, 3 M aq, soln,, pH 7,0, autoclaved
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS No.: 127-09-3
Molecular formula: C₂H₃NaO₂
Formula weight: 82,03
Physical form: liquid
Odour: odourless
MDL No.: MFCD00012459
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together