




Decahydronaphthalene (mixture of cis and trans isomers)
Supplier: Roth Carl
Certificates
About this item
CAS Number:91-17-8
:
Formula:C₁₀H₁₈
MDL Number:MFCD00004130
:138.25 g/mol
Merck Index:13,02866
Boiling Pt:189…191 °C (1013 hPa)
Melting Pt:∼–40 °C
Density:0.8696 g/cm³ (20 °C)
Flash Pt:57 °C
EINECS:202-046-9
UN:1147
ADR:3,III
:GHS02
:GHS06
:GHS05
:GHS08
:GHS09
Storage Temperature:Ambient
Frequently Bought Together