TRIS HCl (tris(hydroxymethyl)aminomethane hydrochloride)
Supplier: EUROCLONE
Certificates
About this item
CAS Number:1185-53-1
:
Formula:NH₂C(CH₂OH)₃·HCl
MDL Number:MFCD00012590
:157.6 g/mol
Merck Index:12,09902
Melting Pt:150…152 °C
Storage Temperature:Ambient
EINECS:214-684-5
Frequently Bought Together