Herbicide reference standard, 2-(2,4,5-Trichlorophenoxy)propionic acid methyl ester 10 µg/ml in ethyl acetate, Dr Ehrenstorfer
Catalog # EHERL13495000EA
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:4841-20-7
- Description:2-(2,4,5-Trichlorophenoxy)propionic acid methyl ester 10 µg/ml in ethyl acetate
- Formula:C₁₀H₉Cl₃O₃
- Description:2-(2,4,5-Trichlorophenoxy)propionic acid methyl ester