Order Entry
Switzerland
ContactUsLinkComponent
Herbicide reference standard, 2-(2,4,5-Trichlorophenoxy)propionic acid 100 µg/ml in dichloromethane, CIL
Herbicide reference standard, 2-(2,4,5-Trichlorophenoxy)propionic acid 100 µg/ml in dichloromethane, CIL
Catalog # PROOCILULM72131.2
CAS Number:  
Herbicide reference standard, 2-(2,4,5-Trichlorophenoxy)propionic acid 100 µg/ml in dichloromethane, CIL
Catalog # PROOCILULM72131.2
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1,2 mL
  • CAS No.:
    93-72-1
  • Description:
    2-(2,4,5-Trichlorophenoxy)propionic acid 100 µg/ml in dichloromethane
  • Formula:
    C₉H₇Cl₃O₃
  • Description:
    2-(2,4,5-Trichlorophenoxy)propionic acid

Specifications

About this item