Supelco® Organic Reference Standard, Geranyl acetate
Catalog # 45896-1ML-F
Supplier: Merck
CAS Number:
Specifications
- Pk:1 mL
- Standards:Analytical standard
- CAS No.:105-87-3
- Description:Geranyl acetate
- Storage conditions:2…8 °C
- Hazard class description:H315 - H317 - H412
- Shipping conditions:STD
- MDL No.:MFCD00015037
- M-Clarity quality level:MQ100
- Synonym(s):trans-3,7-Dimethyl-2,6-octadien-1-yl acetate; trans-3,7-Dimethyl-2,6-octadienyl acetate
- Formula:CH₃CO₂CH₂CH=C(CH₃)CH₂CH₂CH=C(CH₃)₂
- Molecular weight:196,29
- Flash point:104 °C - Closed cup
- Boiling point:138 °C/25 mmHg (lit.)
Specifications
About this item
- GHS signal: GHS07
Caution: P261 - P264 - P272 - P273 - P280 - P302 + P352