Order Entry
Switzerland
ContactUsLinkComponent
Titanium standard dissolved in oil, 5,000 µg/g Ti in mineral oil for wear metal analysis, Specpure®
Titanium standard dissolved in oil, 5,000 µg/g Ti in mineral oil for wear metal analysis, Specpure®
Catalog # 42451.18
CAS Number:  
Titanium standard dissolved in oil, 5,000 µg/g Ti in mineral oil for wear metal analysis, Specpure®
Catalog # 42451.18
CAS Number:  

Specifications

  • Concentration:
    5000 µg/g
  • Matrix:
    Mineral oil
  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g
  • Description:
    Standard for wear metal analysis
  • MDL No.:
    MFCD00011264
  • Element:
    Titanium
  • Symbol:
    Ti

Specifications

About this item

Physical Form: Liquid
MDL No.: MFCD00011264