Calcium standard dissolved in oil, 5,000 µg/g Ca in Base Oil 20
Katalognr. PROOCON150-500-205
Anbieter: LGC Standards PROMOCHEM
CAS-Nummer:
Technische Daten
- Concentration:5000 µg/g
- Matrix:Base Oil 20
- Pack type:Glass bottle for solids
- Pk:50 g
- Environmentally Preferable:
- Element:Calcium
- Symbol:Ca