Vanadium standard dissolved in oil, 1000 µg/g V in mineral oil for wear metal analysis, Specpure®
Supplier: Thermo Fisher Scientific
Certificates
About this item
Physical Form: Liquid
MDL No.: MFCD00011453
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together